Learn More
Biomatik Corporation Recombinant Carica papaya Papain, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20385
The purity of Recombinant Carica papaya Papain is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Carica papaya (Papaya). Alternative Names: Papaya proteinase I. PPI. UniProt: P00784. Expression Region: 134-345aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 27.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4766