Learn More
Biomatik Corporation Recombinant Human Acid ceramidase (ASAH1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20407
The purity of Recombinant Human Acid ceramidase (ASAH1) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: ASAH1. Alternative Names: Acylsphingosine deacylase. N-acylsphingosine amidohydrolase. Putative 32KDA heart protein. PHP32. UniProt: Q13510. Expression Region: 22-395aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 58.7kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4783