Learn More
Biomatik Corporation Recombinant Pseudomonas aeruginosa Regulatory protein RhlR (rhlR), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20478
The purity of Recombinant Pseudomonas aeruginosa Regulatory protein RhlR (rhlR) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Pseudomonas aeruginosa (strain ATCC 15692 / DSM 22644 / CIP 104116 / JCM 14847 / LMG 12228 / 1C / PRS 101 / PAO1). Gene Name: rhlR. Alternative Names: Elastase modulator. UniProt: P54292. Expression Region: 1-241aa. Sequence Info: Full Length. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 47.6kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4830