Learn More
Biomatik Corporation Recombinant Human Matrix Gla protein (MGP), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20060
The purity of Recombinant Human Matrix Gla protein (MGP) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: MGP. Alternative Names: Cell growth-inhibiting gene 36 protein. UniProt: P08493. Expression Region: 20-96aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 13.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4562