Learn More
Biomatik Corporation Recombinant Rat Insulin-like growth factor-binding protein 1 (Igfbp1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20520
The purity of Recombinant Rat Insulin-like growth factor-binding protein 1 (Igfbp1) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Rattus norvegicus (Rat). Gene Name: Igfbp1. UniProt: P21743. Expression Region: 26-272aa. Sequence Info: Full Length of Mature Protein. Tag Info: Tag-Free. Theoretical MW: 26.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4862