Learn More
Biomatik Corporation Recombinant Human 4-aminobutyrate aminotransferase, mitochondrial (ABAT), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20546
The purity of Recombinant Human 4-aminobutyrate aminotransferase, mitochondrial (ABAT) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: ABAT. Alternative Names: (S)-3-amino-2-methylpropionate transaminase (EC: 2.6.1.22). GABA aminotransferase. UniProt: P80404. Expression Region: 29-500aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 73.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4887