Learn More
Biomatik Corporation Recombinant Escherichia coli K99 fimbrial protein (fanC), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20556
The purity of Recombinant Escherichia coli K99 fimbrial protein (fanC) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Escherichia coli. Gene Name: fanC. UniProt: P18103. Expression Region: 23-181aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 32.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4895