Learn More
Biomatik Corporation Recombinant Human Nuclear receptor subfamily 1 group I member 2 (NR1I2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20560
The purity of Recombinant Human Nuclear receptor subfamily 1 group I member 2 (NR1I2) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: NR1I2. Alternative Names: Orphan nuclear receptor PAR1. Orphan nuclear receptor PXR. Pregnane X receptor. Steroid and xenobiotic receptor. UniProt: O75469. Expression Region: 1-434aa. Sequence Info: Full Length. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 69.8kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4898