Learn More
Biomatik Corporation Recombinant Human ATP synthase subunit alpha, mitochondrial (ATP5F1A), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20568
The purity of Recombinant Human ATP synthase subunit alpha, mitochondrial (ATP5F1A) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: ATP5F1A. UniProt: P25705. Expression Region: 44-553aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 71.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4902