Learn More
Biomatik Corporation Recombinant Human Pregnancy zone protein (PZP), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20068
The purity of Recombinant Human Pregnancy zone protein (PZP), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: PZP. Alternative Names: C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6. UniProt: P20742. Expression Region: 472-821aa. Sequence Info: Partial of Isoform 2. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 42.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4569