Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 (mpt64), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20677
The purity of Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 (mpt64) is greater than 85% as determined by SDS-PAGE. Source: Baculovirus. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh). Gene Name: mpt64. Alternative Names: Antigen MPT64. UniProt: P9WIN8. Expression Region: 24-228aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 10xHis-tagged. Theoretical MW: 24.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4931