Learn More
Biomatik Corporation Recombinant Human Aquaporin-1 (AQP1), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20005
The purity of Recombinant Human Aquaporin-1 (AQP1), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: AQP1. Alternative Names: Aquaporin-CHIP; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule. UniProt: P29972. Expression Region: 220-269aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 21.6kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4534