Learn More
Biomatik Corporation Recombinant Human Bone morphogenetic protein 2 (BMP2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20984
The purity of Recombinant Human Bone morphogenetic protein 2 (BMP2) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: BMP2. Alternative Names: Bone morphogenetic protein 2A; BMP-2A. UniProt: P12643. Expression Region: 283-396aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 16.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4961