Learn More
Biomatik Corporation Recombinant Human Uncharacterized protein C21orf2 (C21orf2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21000
The purity of Recombinant Human Uncharacterized protein C21orf2 (C21orf2) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: C21orf2. Alternative Names: C21orf-HUMF09G8.5. Leucine-rich repeat-containing protein 76. YF5/A2. UniProt: O43822. Expression Region: 1-375aa. Sequence Info: Full Length. Tag Info: Tag-Free. Theoretical MW: 40.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4963