Learn More
Biomatik Corporation Recombinant Human Cerebellar degeneration-related protein 2 (CDR2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21078
The purity of Recombinant Human Cerebellar degeneration-related protein 2 (CDR2) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: CDR2 PCD17. Alternative Names: Major Yo paraneoplastic antigen; Paraneoplastic cerebellar degeneration-associated antigen. UniProt: Q01850. Expression Region: 1-454aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 55.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4971