Learn More
Biomatik Corporation Recombinant Pig Growth hormone receptor (GHR), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21374
The purity of Recombinant Pig Growth hormone receptor (GHR), partial is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Sus scrofa (Pig). Gene Name: GHR. Alternative Names: Somatotropin receptor. UniProt: P19756. Expression Region: 19-264aa. Sequence Info: Partial. Tag Info: C-terminal 6xHis-tagged. Theoretical MW: 30.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5003