Learn More
Biomatik Corporation Recombinant Sheep Interleukin-6 (IL6), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21523
The purity of Recombinant Sheep Interleukin-6 (IL6) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Ovis aries (Sheep). Gene Name: IL6. UniProt: P29455. Expression Region: 30-208aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal GST-tagged. Theoretical MW: 47.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5017