Learn More
Biomatik Corporation Recombinant Human Thrombopoietin receptor (MPL), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21680
The purity of Recombinant Human Thrombopoietin receptor (MPL), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: MPL. Alternative Names: Myeloproliferative leukemia protein; Proto-oncogene c-Mpl; CD110. UniProt: P40238. Expression Region: 26-491aa. Sequence Info: Extracellular Domain. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 56.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5033