Learn More
Biomatik Corporation Recombinant Human Metallothionein-3 (MT3), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21697
The purity of Recombinant Human Metallothionein-3 (MT3) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: MT3. Alternative Names: GIFB. UniProt: P25713. Expression Region: 1-68aa. Sequence Info: Full Length. Tag Info: N-terminal GST-tagged. Theoretical MW: 33.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5034