Learn More
Biomatik Corporation Recombinant Escherichia coli Peptide deformylase (def), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21798
The purity of Recombinant Escherichia coli Peptide deformylase (def) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Escherichia coli (strain K12). Gene Name: def. Alternative Names: Polypeptide deformylase. UniProt: P0A6K3. Expression Region: 2-169aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 35.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5051