Learn More
Biomatik Corporation Recombinant Human Phosphoribosylformylglycinamidine synthase (PFAS), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC21807
The purity of Recombinant Human Phosphoribosylformylglycinamidine synthase (PFAS), partial is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: PFAS. UniProt: O15067. Expression Region: 1064-1302aa. Sequence Info: Partial. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 45.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5055