Learn More
Biomatik Corporation Recombinant Rat Transforming growth factor beta-1 (Tgfb1), partial, 100UG
Supplier:  Biomatik Corporation RPC22107
Encompass_Preferred
The purity of Recombinant Rat Transforming growth factor beta-1 (Tgfb1), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Rattus norvegicus (Rat). Gene Name: Tgfb1. UniProt: P17246. Expression Region: 30-278aa. Sequence Info: Partial. Tag Info: Tag-Free. Theoretical MW: 28.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5088