Learn More
Biomatik Corporation Recombinant Rat Tryptase (Tpsab1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22164
The purity of Recombinant Rat Tryptase (Tpsab1) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Rattus norvegicus (Rat). Gene Name: Tpsab1. Alternative Names: Mast cell protease 7. UniProt: P27435. Expression Region: 29-273aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 47.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5092