Learn More
Biomatik Corporation Recombinant Human Transthyretin (TTR), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22183
The purity of Recombinant Human Transthyretin (TTR) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: TTR. Alternative Names: ATTRPrealbumin; TBPA. UniProt: P02766. Expression Region: 21-147aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 17.8kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5096