Learn More
Biomatik Corporation Recombinant Human Elongation factor Tu, mitochondrial (TUFM), 100UG
Supplier:  Biomatik Corporation RPC22192
Encompass_Preferred
The purity of Recombinant Human Elongation factor Tu, mitochondrial (TUFM) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: TUFM. Alternative Names: P43. UniProt: P49411. Expression Region: 44-452aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 49kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5100