Learn More
Biomatik Corporation Recombinant Human Versican core protein (VCAN), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22218
The purity of Recombinant Human Versican core protein (VCAN), partial is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: VCAN. Alternative Names: Chondroitin sulfate proteoglycan core protein 2. UniProt: P13611. Expression Region: 3089-3354aa. Sequence Info: Partial. Tag Info: N-terminal 10xHis-B2M-tagged and C-terminal Myc-tagged. Theoretical MW: 47.6kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5103