Learn More
Biomatik Corporation Recombinant Danio rerio Interleukin 1,beta (il1b), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22289
The purity of Recombinant Danio rerio Interleukin 1,beta (il1b) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Danio rerio (Zebrafish) (Brachydanio rerio). Gene Name: il1b. UniProt: E6N152. Expression Region: 41-272aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 29.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5109