Learn More
Biomatik Corporation Recombinant Hypocrea jecorina Hydrophobin-2 (hfb2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22329
The purity of Recombinant Hypocrea jecorina Hydrophobin-2 (hfb2) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Hypocrea jecorina (Trichoderma reesei). Gene Name: hfb2. Alternative Names: Hydrophobin II. UniProt: P79073. Expression Region: 16-86aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 10xHis-B2M-JD-tagged. Theoretical MW: 12.7kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5112