Learn More
Biomatik Corporation Recombinant Human Complement C4-B (C4B),partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22377
The purity of Recombinant Human Complement C4-B (C4B), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: C4B. Alternative Names: Basic complement C4C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3. UniProt: P0C0L5. Expression Region: 1454-1744aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 49.1kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5117