Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis Diacylglycerol acyltransferase/mycolyltransferase Ag85B (fbpB), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22384
The purity of Recombinant Mycobacterium tuberculosis Diacylglycerol acyltransferase/mycolyltransferase Ag85B (fbpB) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh). Gene Name: fbpB. Alternative Names: 30KDA extracellular protein. Acyl-CoA: diacylglycerol acyltransferase. Antigen 85 complex B. UniProt: P9WQP0. Expression Region: 41-325aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 36.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5118