Learn More
Biomatik Corporation Recombinant Absidia glauca Actin-1 (ACT1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22426
The purity of Recombinant Absidia glauca Actin-1 (ACT1) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Absidia glauca (Pin mould). Gene Name: ACT1. UniProt: P10982. Expression Region: 1-140aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged and C-terminal Myc-tagged. Theoretical MW: 21.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5124