Learn More
Biomatik Corporation Recombinant Pig Vascular endothelial growth factor A (VEGFA), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20101
The purity of Recombinant Pig Vascular endothelial growth factor A (VEGFA) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Sus scrofa (Pig). Gene Name: VEGFA. Alternative Names: Vascular permeability factor; VPF. UniProt: P49151. Expression Region: 27-190aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 23.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4592