Learn More
Biomatik Corporation Recombinant Parietaria judaica Probable non-specific lipid-transfer protein 2 (LTP2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22622
The purity of Recombinant Parietaria judaica Probable non-specific lipid-transfer protein 2 (LTP2) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Parietaria judaica (Pellitory-of-the-wall) (Parietaria diffusa). Gene Name: LTP2. Alternative Names: Allergen Par j II. Major pollen allergen Par j 2.0101. Protein P2. Allergen: Par j 2.0101. UniProt: P55958. Expression Region: 32-133aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 27.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5160