Learn More
Biomatik Corporation Recombinant Pseudomonas aeruginosa Hemolytic phospholipase C (plcH), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22769
The purity of Recombinant Pseudomonas aeruginosa Hemolytic phospholipase C (plcH), partial is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Pseudomonas aeruginosa (strain ATCC 15692 / DSM 22644 / CIP 104116 / JCM 14847 / LMG). Gene Name: plcH. Alternative Names: Heat-labile hemolysin. PLC-H. Phosphatidylcholine cholinephosphohydrolase. UniProt: P06200. Expression Region: 51-436aa. Sequence Info: Partial. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 64.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5185