Learn More
Biomatik Corporation Recombinant Human papillomavirus type 18 Protein E7 (E7), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22775
The purity of Recombinant Human papillomavirus type 18 Protein E7 (E7) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Human papillomavirus type 18. Gene Name: E7. UniProt: P06788. Expression Region: 1-105aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 16kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5186