Learn More
Biomatik Corporation Recombinant Human Frataxin, mitochondrial (FxN), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC22972
The purity of Recombinant Human Frataxin, mitochondrial (FxN) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: FxN. Alternative Names: Friedreich ataxia protein. UniProt: Q16595. Expression Region: 1-210aa. Sequence Info: Full Length. Tag Info: N-terminal 10xHis-tagged and C-terminal Myc-tagged. Theoretical MW: 28.1kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5218