Learn More
Biomatik Corporation Recombinant Phleum pratense Pollen allergen Phl p 5a, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23095
The purity of Recombinant Phleum pratense Pollen allergen Phl p 5a is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Phleum pratense (Common timothy). UniProt: Q40962. Expression Region: 1-286aa. Sequence Info: Full Length. Tag Info: N-terminal 10xHis-SUMO-tagged and C-terminal Myc-tagged. Theoretical MW: 48.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5231