Learn More
Biomatik Corporation Recombinant Colwellia psychrerythraea DNA polymerase IV (dinB), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23108
The purity of Recombinant Colwellia psychrerythraea DNA polymerase IV (dinB) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Colwellia psychrerythraea (strain 34H / ATCC BAA-681) (Vibrio psychroerythus). Gene Name: dinB. UniProt: Q487H6. Expression Region: 1-352aa. Sequence Info: Full Length. Tag Info: Tag-Free. Theoretical MW: 39.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5237