Learn More
Biomatik Corporation Recombinant Mouse RNA polymerase II subunit A C-terminal domain phosphatase (Ctdp1), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23208
The purity of Recombinant Mouse RNA polymerase II subunit A C-terminal domain phosphatase (Ctdp1), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Ctdp1. Alternative Names: TFIIF-associating CTD phosphatase. UniProt: Q7TSG2. Expression Region: 178-341aa. Sequence Info: Partial. Tag Info: N-terminal 10xHis-tagged and C-terminal Myc-tagged. Theoretical MW: 24kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5247