Learn More
Biomatik Corporation Recombinant Mouse Plexin domain-containing protein 1 (Plxdc1), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23373
The purity of Recombinant Mouse Plexin domain-containing protein 1 (Plxdc1), partial is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Plxdc1. Alternative Names: Tumor endothelial marker 7. Tem7. UniProt: Q91ZV7. Expression Region: 20-426aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 49.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5268