Learn More
Biomatik Corporation Recombinant Mouse [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial (Pdk3), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23469
The purity of Recombinant Mouse [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial (Pdk3), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Pdk3. Alternative Names: Pyruvate dehydrogenase kinase isoform 3. UniProt: Q922H2. Expression Region: 131-362aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 30.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5277