Learn More
Biomatik Corporation Recombinant Human SH3 domain-containing YSC84-like protein 1 (SH3YL1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23475
The purity of Recombinant Human SH3 domain-containing YSC84-like protein 1 (SH3YL1) is greater than 85% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: SH3YL1. UniProt: Q96HL8. Expression Region: 1-342aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 41.1kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5278