Learn More
Biomatik Corporation Recombinant Mouse 14-3-3 protein beta/alpha (Ywhab), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23635
The purity of Recombinant Mouse 14-3-3 protein beta/alpha (Ywhab) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Ywhab. Alternative Names: Protein kinase C inhibitor protein 1. UniProt: Q9CQV8. Expression Region: 1-246aa. Sequence Info: Full Length. Tag Info: N-terminal 10xHis-tagged. Theoretical MW: 33.6kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5290