Learn More
Biomatik Corporation Recombinant Rat Gremlin-1 (Grem1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC23825
The purity of Recombinant Rat Gremlin-1 (Grem1) is greater than 85% as determined by SDS-PAGE. Source: Mammalian cell. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Rattus norvegicus (Rat). Gene Name: Grem1. Alternative Names: Cysteine knot superfamily 1, BMP antagonist 1. Down-regulated in Mos-transformed cells protein. Cktsf1b1, Drm. UniProt: O35793. Expression Region: 25-184aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 10xHis-tagged and C-terminal Myc-tagged. Theoretical MW: 22.2kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5299