Learn More
Biomatik Corporation Recombinant Human Clusterin (CLU), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC24107
The purity of Recombinant Human Clusterin (CLU), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: CLU. Alternative Names: Aging-associated gene 4 protein; Apolipoprotein J; Apo-JComplement cytolysis inhibitor; CLIComplement-associated protein SP-40,40Ku70-binding protein 1NA1/NA2Testosterone-repressed prostate message 2; TRPM-2. UniProt: P10909. Expression Region: 23-224aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 27.8kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5319