Learn More
Biomatik Corporation Recombinant Human Interleukin-15 (IL15), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC24129
The purity of Recombinant Human Interleukin-15 (IL15) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: IL15. UniProt: P40933. Expression Region: 49-162aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 16.8kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5320