Learn More
Biomatik Corporation Recombinant Human Protein-lysine 6-oxidase (LOX), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC24401
The purity of Recombinant Human Protein-lysine 6-oxidase (LOX), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: LOX. Alternative Names: Lysyl oxidase. UniProt: P28300. Expression Region: 174-417aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 32.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5334