Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Exfoliative toxin A (eta), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20131
The purity of Recombinant Staphylococcus aureus Exfoliative toxin A (eta) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Staphylococcus aureus. Gene Name: eta. Alternative Names: Epidermolytic toxin A. UniProt: P09331. Expression Region: 39-280aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 30.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4614