Learn More
Biomatik Corporation Recombinant Mouse Osteopontin (Spp1), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC24965
The purity of Recombinant Mouse Osteopontin (Spp1), partial is greater than 90% as determined by SDS-PAGE. Source: Yeast. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Spp1. Alternative Names: 2ARBone sialoprotein 1Calcium oxalate crystal growth inhibitor protein; Early T-lymphocyte activation 1 protein; Minopontin; Secreted phosphoprotein 1; SPP-1. UniProt: P10923. Expression Region: 17-290aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 32.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5379