Learn More
Biomatik Corporation Recombinant Human Mixed lineage kinase domain-like protein (MLKL), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC25495
The purity of Recombinant Human Mixed lineage kinase domain-like protein (MLKL) is greater than 90% as determined by SDS-PAGE. Source: Yeast. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: MLKL. UniProt: Q8NB16. Expression Region: 1-471aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 56.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5441